Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Borohydride Solution

Catalog No. 230200800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-020-0800ML 800 mL
23-020-0100ML 100 mL
23-020-025ML 4 x 25 mL
23-020-02L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 23-020-0800ML Supplier Sigma Aldrich Supplier No. 230200800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of gallium, indium, rhenium and zinc tris(mercaptoimidazolyl)hydroborato complexesMechano-chemical metathesis reactionsNoncatalytic hydrolysis for hydrogen generationGrowth of large gold monolayer protected-clustersAnion substitution reactionsDehydrogenation reactionsSure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 16949-15-8
Quantity 800 mL
Molecular Formula H4BLi
Linear Formula LiBH4
MDL Number MFCD00011088
Molecular Weight (g/mol) 21.78 g/mol
Density 0.896 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.