Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lithium bromide, anhydrous, 99% min

Catalog No. AA134080E
Change view
Click to view available options
Quantity:
2500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA134080E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA134080E Supplier Thermo Scientific™ Supplier No. 134080E
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.