Learn More
Lithium Bromide, MP Biomedicals™
Lithium Bromide is a hygroscopic chemical.
Supplier: MP Biomedicals, Inc 0215156180
| Quantity | 100 g |
|---|
Description
- Appearance: White powder
- Molecular Formula: LiBr
- Molecular Weight: 86.844 g/mol
- CAS Number: 7550-35-8
- Hygroscopic chemical
- Storage Temperature: 15°C to 30°C
- For research or further manufacturing use only
Chemical Identifiers
| 7550-35-8 | |
| 86.84 | |
| lithium(1+) bromide |
| BrLi | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| [Li+].[Br-] |
Specifications
| White | |
| Powder | |
| BrLi | |
| MFCD00011077 | |
| Soluble in water | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| lithium(1+) bromide | |
| 26038 | |
| 86.844 g/mol | |
| USA | |
| White Powder |
| 7550-35-8 | |
| 100 g | |
| LiBr | |
| lithium(1+) bromide | |
| InChI=1S/BrH.Li/h1H;/q;+1/p-1 | |
| [Li+].[Br-] | |
| 86.84 | |
| CHEBI:32662 | |
| Research | |
| Hygroscopic | |
| Lithium Bromide |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.