Learn More
Lithium Bromide, MP Biomedicals™
Lithium Bromide is a hygroscopic chemical.
$105.00 - $812.00
Chemical Identifiers
| CAS | 7550-35-8 |
|---|---|
| Molecular Formula | BrLi |
| Molecular Weight (g/mol) | 86.84 |
| InChI Key | AMXOYNBUYSYVKV-UHFFFAOYSA-M |
| IUPAC Name | lithium(1+) bromide |
| SMILES | [Li+].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
02-151-56180
|
MP Biomedicals, Inc
0215156180 |
100 g |
Each for $105.00
|
|
|||||
|
02-151-56190
|
MP Biomedicals, Inc
0215156190 |
500 g |
Each for $430.00
|
|
|||||
|
02-151-56191
|
MP Biomedicals, Inc
0215156191 |
1 kg |
Each for $812.00
|
|
|||||
Description
Chemical Identifiers
| 7550-35-8 | |
| 86.84 | |
| lithium(1+) bromide |
| BrLi | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| [Li+].[Br-] |
Specifications
| White | |
| Powder | |
| LiBr | |
| lithium(1+) bromide | |
| InChI=1S/BrH.Li/h1H;/q;+1/p-1 | |
| [Li+].[Br-] | |
| 86.84 | |
| CHEBI:32662 | |
| Research | |
| Hygroscopic | |
| Lithium Bromide |
| 7550-35-8 | |
| BrLi | |
| MFCD00011077 | |
| Soluble in water | |
| AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
| lithium(1+) bromide | |
| 26038 | |
| 86.844 g/mol | |
| USA | |
| White Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.