Learn More
Description
- Appearance: White powder
- Molecular Formula: LiBr
- Molecular Weight: 86.844 g/mol
- CAS Number: 7550-35-8
- Hygroscopic chemical
- Storage Temperature: 15°C to 30°C
- For research or further manufacturing use only
- Chemical formula: BrLi
- Physical form: Powder
Specifications
Specifications
| Boiling Point | 1265°C |
| Melting Point | 550°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Linear Formula | LiBr |
| MDL Number | MFCD00003503 |
| Synonym | lithium(1+) bromide |
| Solubility Information | Soluble in water |
| InChI Formula | InChI=1S/BrH.Li/h1H;/q;+1/p-1 |
| Show More |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.