Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, ultra dry, 99.998% (metals basis)

Catalog No. AA4419906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4419906 5 g
AA4419914 25 g
AA4419922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4419906 Supplier Thermo Scientific Chemicals Supplier No. 04419906
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7550-35-8 | BrLi | 86.844 g/mol

Employed as adsorbents, functional fluids (closed systems) and intermediates. Used as an operating medium for air-conditioning and industrial drying system. It is also used in manufacturing pharmaceuticals and alkylation process. It is used as brazing and welding fluxes. Used for reconstitution of brines, as swelling agent for proteins, as electrolyte component in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265°C
Melting Point 550°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Soluble in water (at 20°C 1490g/L),alcohol and glycol.
Formula Weight 86.85
Sensitivity Hygroscopic
Density 3.464
Chemical Name or Material Lithium bromide, ultra dry
Show More Show Less
Hazard Category H302-H315-H317-H319
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 231-439-8
RTECSNumber OJ5755000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.