Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide, ultra dry, 99.998% (metals basis)

Catalog No. AA1453722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1453722 100 g
AA1453706 5 g
AA1453714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1453722 Supplier Thermo Scientific Chemicals Supplier No. 01453722
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7550-35-8 | BrLi | 86.844 g/mol

Lithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265°C
Melting Point 550°C
Color White
Physical Form Beads
Quantity 100 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Very soluble in water and alcohol. Soluble in methanol,acetone,and glycol. Slightly soluble in pyridine.
Formula Weight 86.85
Sensitivity Hygroscopic
Density 3.464
Chemical Name or Material Lithium bromide
Show More Show Less
Hazard Category H302-H315-H317-H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 231-439-8
RTECSNumber OJ5755000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.