Learn More
Justrite Lithium Cabinet
GREENER_CHOICE
Catalog No. 18821454 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
18-821-454 Gray
1 options
Catalog No. 18-821-454 Supplier Justrite Supplier No. 231703
  • $5,411.00
    $3,888.65 / Each of 1
    Save $1,522.35
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
Justrite's lithium-ion battery charging and storage cabinet, fortified with ChargeGuard™ for ultimate protection.
  • Ideal for charging and temporary storage of lithium-ion batteries
  • 2 kWh TECR maximum total capacity
  • Includes 8-receptacle power strip
  • Heat-reactive label changes colors when external temperatures reach 120°F
  • Shelf capacity: 65 lb. of evenly distributed weight
  • Cabinet Exterior Dimensions: 24 x 43 x 18 in.
  • Required Utilities: This product is intended to be plugged into a dedicated 120VAC/60Hz GFCI supply using a minimum 14 gauge cord. (Supply cord not included with cabinet)
Specifications
Door Style Double, Self-close, Three-point lock, Hinged, Locking
Color Gray
Material Steel (18 ga), Double-wall Steel
Product Type Flammables Storage Cabinet