Learn More
Alkali Scientific Lithium Chloride, 1 Kg
SDP
Supplier:  Alkali Scientific LIT002CP
Encompass_Preferred
Lithium Chloride, 1 kg, is a high-purity laboratory-grade reagent used in chemical and biochemical applications. This larger 1 kg size is designed for high-volume procedures that require precise, consistent lithium chloride. It is commonly used in lithium-based reactions, as a drying agent, and for preparing lithium salts in organic synthesis. Lithium chloride is also involved in processes that require high-purity lithium compounds, such as the production of lithium batteries and pharmaceutical intermediates. This product is well-suited for laboratories requiring significant quantities of high-purity lithium chloride for research and production.
Catalog No. 50-006-60137