Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, 1 Molar in Glacial Acetic Acid, Reagents

Catalog No. 01334827
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-827 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-827 Supplier Reagents Holdings Llc Supplier No. CL197500500A
Add to Cart
Add to Cart

Lithium Chloride, 1 Molar in Glacial Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

  • Grade: Reagent
  • Physical form: Liquid
  • Color: Colorless
  • pH: <2
  • Boiling point: 118°C
  • Specific gravity: 1.0705
  • Packaging: Poly bottle
  • Manufactured in an ISO 9001 facility

Chemical Identifiers

CAS 64-19-7 , 7447-41-8
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Boiling Point 118°C
Melting Point 16°C to 17°C
Color Colorless
pH <2
Physical Form Liquid
Quantity 500 mL
Linear Formula LiCl, CH3COOH
UN Number UN2789
Flash Point 39°C (102°F)
InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
Formula Weight 60.05
Grade Reagent
Concentration or Composition (by Analyte or Components) 1 M Lithium Chloride in Glacial Acetic Acid
Manufacturing Origin USA
Ignition Point 465°C
Packaging Poly Bottle
Appearance Colorless Liquid
Specific Gravity 1.0705
Decomposition Information Carbon Oxides
Chemical Name or Material Lithium Chloride, 1 Molar in Glacial Acetic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.