Learn More
Alkali Scientific Lithium Chloride, 500 G
SDP
Supplier:  Alkali Scientific LIT003CP
Encompass_Preferred
Lithium Chloride, 500g, is a laboratory-grade reagent commonly used in organic synthesis and various chemical processes. Lithium chloride is essential in the preparation of lithium-based compounds, which are widely used in research and industrial applications, including the production of lithium batteries. It is also used as a drying agent for organic materials and as a component in the synthesis of other lithium salts. The 500g size is convenient for moderate-scale laboratory experiments and procedures requiring high-purity lithium chloride for accurate and reproducible results.
Catalog No. 50-006-60547