Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lithium Chloride 8M, OmniPur™, Calbiochem™,

Catalog No. 5922500ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-225-00ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-225-00ML Supplier MilliporeSigma™ Supplier No. 5922500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for precipitation of RNA prior to PCR or cDNA synthesis

Separates RNA from inhibitors of cDNA synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Linear Formula LiCl
Solubility Information Soluble in water
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.39
Grade Reagent
Concentration or Composition (by Analyte or Components) 7.7 to 8.2M
Product Line OmniPur
Appearance Clear, Colorless Liquid
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.