Learn More
Strem, An Ascensus Company CAS# 7447-41-8. 250g. Lithium chloride, 99% (ACS). MFCD00011078
SDP
Catalog No. 5090113602
Encompass
CAS# 7447-41-8. 250g. Lithium chloride, 99% (ACS). MFCD00011078. Molecular Weight: 42.39. Molecular Formula: LiCl. Color/form: white pwdr. Strem# 93-0313. http://www.strem.com/catalog/v/93-0313/
Catalog No. 50-901-13602 Supplier Strem, An Ascensus Company Supplier No. 930313250G
CAS# 7447-41-8. 250g. Lithium chloride, 99% (ACS). MFCD00011078. Molecular Weight: 42.39. Molecular Formula: LiCl. Color/form: white pwdr. Strem# 93-0313. http://www.strem.com/catalog/v/93-0313/