Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, 99+%, ACS reagent

Catalog No. AC413270025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413270025 2.5 kg Plastic bottle
AC413270050 5 g Glass Bottle
AC413271000 100 g Plastic bottle
AC413275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC413270025 Supplier Thermo Scientific Chemicals Supplier No. 413270025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 7447-41-8 | ClLi | 42.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382.0°C
Melting Point 605.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15, 5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
IUPAC Name lithium;chloride
Formula Weight 42.38
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 1.0% max. (1 g, 105°C, 2 hrs)
Packaging Plastic bottle
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride, 99%
Show More Show Less
What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.

Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse
EINECSNumber 231-212-3
RTECSNumber OJ5950000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.