Learn More
Lithium chloride, 99+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1382.0°C |
| Melting Point | 605.0°C |
| Color | White |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 500 g |
| Loss on Drying | 1.0% max. (1 g, 105°C, 2 hrs) |
| Packaging | Plastic bottle |
| Specific Gravity | 2.06 |
| Assay Percent Range | 99% min. (Argentometry) |
| Density | 2.0600g/mL |
| Show More |
Frequently Asked Questions (FAQs)
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse |
| EINECSNumber | 231-212-3 |
| RTECSNumber | OJ5950000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.