Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, ≥99%, MP Biomedicals

Catalog No. ICN19401090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19401090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19401090 Supplier MP Biomedicals, Inc Supplier No. 0219401090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity lithium chloride reagent for general laboratory use.

  • Assay: ≥99%
  • Molecular Formula: ClLi
  • Formula Weight: 42.39 g/mol
  • CAS Number: 7447-41-8
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 2417 to 2480°F
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range ≥99%
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Soluble in water
Formula Weight 42.4
Percent Purity ≥99%
Packaging Bottle
Appearance White Crystalline Powder
Decomposition Information Hydrogen Chloride, Lithium Oxide
Chemical Name or Material Lithium Chloride
Show More Show Less
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.