Learn More
Description
- Chemical Name: Lithium Chloride
- Grade: ACS
- Physical Form: Granular
- Formula Weight: 42.39
- Molecular Formula: ClLi
- CAS Number: 7447-41-8
Specifications
Specifications
| Color | White |
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Linear Formula | LiCl |
| Solubility Information | Soluble in water |
| Formula Weight | 42.39 |
| Grade | Meets ACS Reagent Chemical Requirements |
| Odor | Odorless |
| Packaging | Poly |
| Product Line | Macron Fine Chemicals |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.