Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, ACS Reagent, Solstice

Catalog No. 6002283 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-83 100 g
60-022-85 500 g
60-022-84 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-022-83 Supplier Solstice Supplier No. 310468100G
Add to Cart
Add to Cart

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium;chloride
SMILES [Li+].[Cl-]

Specifications

Melting Point 605°C
Color White
pH 6
Quantity 100 g
Assay Percent Range ≥99%
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Highly soluble in water
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.39g/mol
Percent Purity ≥99%
Grade Meets ACS reagent chemical requirements
Manufacturing Origin USA
Identification Passes Test
Packaging Plastic Bottle
Product Line Solstice
Sensitivity Hygroscopic
Appearance White Crystalline Powder
Chemical Name or Material Lithium Chloride
Show More Show Less
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-212-3
RTECSNumber OJ5950000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.