Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-212 2.5 kg Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-212 Supplier Avantor Supplier No. 237005
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Physical Form Granules
Quantity 2.5 kg
Assay Percent Range 99.00%
Linear Formula LiCl
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.39
Percent Purity ≥99.0%
Grade Meets ACS reagent chemical requirements
Manufacturing Origin Germany
Loss on Drying max. 1.0%
Packaging Wide Mouth Amber Glass Bottle
Sensitivity Hygroscopic
Appearance White Granules
Specific Gravity 1L = 2.07kg
Assay by Ag Titrn
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.