Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603925 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
125 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-925 25 g Amber Glass Bottle
18-603-926 125 g Amber Glass Bottle
18-603-928 500 g Amber Glass Bottle
18-603-927 6 x 125 g Amber Glass Bottle
18-603-930 2.5 kg Amber Glass Bottle
18-603-929 6 x 500 g Amber Glass Bottle
18-603-931 12 kg Poly Pail
18-603-932 125 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-603-925 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L110925GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

L1109, 7447-41-8, LiCl

Spectrum™ Chemical Lithium Chloride, Crystal, Reagent, ACS is a chemical compound that is extremely soluble in polar solvents and is used in order to obtain lithium metal and as a synthetic reagent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Melting Point 613°C
CAS Max % 100%
Quantity 25 g
Assay Percent Range 99%
Linear Formula LiCl
UN Number NONH for all modes of transport
Synonym lithium chloride, lithium(1+) chloride
Solubility Information Extremely soluble in polar solvents
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
PubChem CID 433294
Formula Weight 42.39
Percent Purity 99%
Grade ACS
Manufacturing Origin USA
Loss on Drying 1%
Odor Odorless
Packaging Amber Glass Bottle
Sensitivity Deliquescent
Appearance White crystalline powder
Decomposition Information Hydrogen Chloride, Lithium Oxides
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.