Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603932 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
125 lb.
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-932 125 lb. Amber Glass Bottle
18-603-925 25 g Amber Glass Bottle
18-603-926 125 g Amber Glass Bottle
18-603-928 500 g Amber Glass Bottle
18-603-927 6 x 125 g Amber Glass Bottle
18-603-930 2.5 kg Amber Glass Bottle
18-603-929 6 x 500 g Amber Glass Bottle
18-603-931 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-603-932 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1109125LBBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

L1109, 7447-41-8, LiCl

Spectrum™ Chemical Lithium Chloride, Crystal, Reagent, ACS is a chemical compound that is extremely soluble in polar solvents and is used in order to obtain lithium metal and as a synthetic reagent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Color White
CAS Max % 100%
Physical Form Crystals or Needles
Quantity 125 lb.
Assay Percent Range 99%
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Extremely soluble in polar solvents
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
PubChem CID 433294
Formula Weight 42.39
Percent Purity 99%
Grade ACS/Reagent
Manufacturing Origin USA
Ignition Point 430°C
Loss on Drying 1%
Odor Odorless
Packaging Amber Glass Bottle
Product Line Spectrum
Sensitivity Deliquescent
Decomposition Information Hydrogen Chloride, Lithium Oxides
Chemical Name or Material Lithium Chloride, Crystal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.