Learn More
Selleck Chemical LLC Lithium Chloride-E0153-1G
SDP
Catalog No. 503136794
Encompass_Preferred
Lithium Chloride (LiCl) inhibits the replication of type 1 and type 2 Herpes simplex virus at concentrations which permitted host cell replication Lithium chloride also prevents glucocorticoid-induced osteonecrosis of femoral heads and strengthens mesenchymal stem cell activity in rats Lithium Chloride can be used to induce animal models of Chronic Epilepsy
Catalog No. 50-313-6794 Supplier Selleck Chemical LLC Supplier No. E01531G
Lithium Chloride (LiCl) inhibits the replication of type 1 and type 2 Herpes simplex virus at concentrations which permitted host cell replication Lithium chloride also prevents glucocorticoid-induced osteonecrosis of femoral heads and strengthens mesenchymal stem cell activity in rats Lithium Chloride can be used to induce animal models of Chronic Epilepsy