Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%, Solstice

Catalog No. 60047518 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-518 6 x 500 g Poly Bottle
60-047-517 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-518 Supplier Solstice Supplier No. 7464606X500G
Add to Cart
Add to Cart

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium;chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1,360°C
Melting Point 605°C (literature)
CAS Max % ≤100.0000%
Physical Form Anhydrous
Quantity 6 x 500 g
Linear Formula LiCl
Formula Weight 42.39
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.07 g/cm3
Chemical Name or Material Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
ShelfLife 720 days from date of manufacture
EINECSNumber 231-212-3
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.