Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003213 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-213 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-213 Supplier Avantor Supplier No. 237001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

  • Meets ACS Reagent Chemical Requirements
  • Grade: BAKER ANALYZED™ A.C.S. Reagent
  • Physical form: Granular
  • Assay (LiCl) (by Ag titrn): 99.6 %
  • Insoluble matter: < 0.01 %
  • Loss on drying: 0.1 %/

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Physical Form Granules
Quantity 500 g
Assay Percent Range 99.6%
Linear Formula LiCl
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.39
Percent Purity ≥99.6%
Grade Meets ACS Reagent Chemical requirements
Manufacturing Origin Germany
Loss on Drying 0.1%
Packaging Poly Bottle
Product Line J.T. Baker
Sensitivity Hygroscopic
Specific Gravity 1L = 2.07kg
Assay by Ag Titrn
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.