Learn More
Medchemexpress LLC Lithium chloride hyd 100mg
SDP
Catalog No. 5000206227
Encompass_Preferred
Lithium chloride hydrate an orally active mood stabilizer is a potent virus inhibitor and effective immunomodulatory agent Lithium chloride hydrate has antidepressant activity by inhibiting GSK3 and promoting neurogenesis Lithium chloride hydrate alleviates cognition dysfunction and the symptoms of acute mania and depression Lithium chloride hydrate can also be used for research of virus infection and Alzheimer s disease[1][2][3]
Catalog No. 50-002-06227 Supplier Medchemexpress LLC Supplier No. HYW094474100MG
Lithium chloride hydrate an orally active mood stabilizer is a potent virus inhibitor and effective immunomodulatory agent Lithium chloride hydrate has antidepressant activity by inhibiting GSK3 and promoting neurogenesis Lithium chloride hydrate alleviates cognition dysfunction and the symptoms of acute mania and depression Lithium chloride hydrate can also be used for research of virus infection and Alzheimer s disease[1][2][3]