Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride Molecular Biology Reagent MP Biomedicals

Catalog No. ICN19401080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19401080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19401080 Supplier MP Biomedicals, Inc Supplier No. 0219401080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity molecular biology grade lithium chloride, optimized for sensitive research applications.

  • CAS Number: 7447-41-8
  • Molecular Formula: LiCl
  • Molecular Weight: 42.39
  • Purity: 99.7%
  • Physical State: White solid
  • Solubility: Soluble in water
  • Boiling Point: 1360°C
  • Storage Conditions: 15°C to 30°C, desiccate
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1360°C
Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range 99%+
Linear Formula LiCl
Solubility Information Soluble in water
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.4
Percent Purity >99%
Grade Molecular Biology
Manufacturing Origin USA
Identification Passes Test
Sensitivity Hygroscopic
Appearance White Powder
Density 2.1g/mL
Chemical Name or Material Lithium Chloride, Molecular Biology Reagent
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.