Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride monohydrate, 99.95% (metals basis)

Catalog No. p-4824796
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2215114 25 g
AA2215122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2215114 Supplier Thermo Scientific Chemicals Supplier No. 02215114
Only null left
Add to Cart

CAS: 16712-20-2 | ClH2LiO | 60.405 g/mol

Lithium chloride monohydrate is used as a precursor in the production of lithium metal and other lithium compounds. It acts as a welding agent for aluminum, salt bath for heat -treatment by low temperature and soldering techniques. It is also used as a tracer for chemical products viz. denaturation of wine. For absorbers, it is an absorption and disinfection reagent as well as desiccant for air conditioner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16712-20-2
Molecular Formula ClH2LiO
Molecular Weight (g/mol) 60.405
MDL Number MFCD00011078
InChI Key VXJIMUZIBHBWBV-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
IUPAC Name lithium;chloride;hydrate
SMILES [Li+].O.[Cl-]

Specifications

Melting Point >98°C-H2O
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 60.41 (42.39 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.78
Chemical Name or Material Lithium chloride monohydrate
Show More Show Less
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 231-212-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.