Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride-potassium chloride

Catalog No. AA1820618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1820618 50 g
AA1820630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1820618 Supplier Thermo Scientific Chemicals Supplier No. 01820618
Add to Cart

CAS: 65567-96-6 | Cl2KLi | 116.938 g/mol

Lithium chloride-potassium chloride is used in separators for mixed transition-metal oxide cathodes in thermal batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65567-96-6
Molecular Formula Cl2KLi
Molecular Weight (g/mol) 116.938
MDL Number MFCD00144623
InChI Key HCQWRNRRURULEY-UHFFFAOYSA-L
Synonym lithium chloride-potassium chloride, acmc-20akmw, lithium potassium chloride, lithium potassium chloride 1/1/2, lithium chloride/potassium chloride eutectic, lithium 1+ ion potassium dichloride, lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis
PubChem CID 16213218
IUPAC Name lithium;potassium;dichloride
SMILES [Li+].[Cl-].[Cl-].[K+]

Specifications

Melting Point 357°C
Color White
Physical Form Crystals
Quantity 50 g
Solubility Information Soluble in water.
Odor Odorless
Sensitivity Hygroscopic
Density 2.02
Chemical Name or Material Lithium chloride-potassium chloride
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.