Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride Solution, 1 M in Ethanol, MilliporeSigma™ Supelco™

Catalog No. 111018950 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8950 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8950 Supplier MilliporeSigma Supelco Supplier No. 62481250MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lithium chloride solution is generally used as a liquid desiccant for hybrid solar desiccant cooling systems. Though it is corrosive in nature, it does not vaporize in air at ambient conditions.

  • Composition: 1 M Lithium chloride in ethanol
  • Molecular weight: 42.39 g/mol
  • Physical state: Liquid
  • Density: 0.83 g/cm³
  • Flash point: 14°C
  • Applications: Generally used as a liquid desiccant for hybrid solar desiccant cooling systems
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Chloride
CAS 7447-41-8
Physical Form Liquid
Quantity 250 mL
Linear Formula LiCl
MDL Number MFCD00011078
UN Number UN1170
Flash Point 14°C (57.2°F)
Synonym LiCl, lithium chloride
IUPAC Name ethanol lithium chloride
Molecular Weight (g/mol) 42.39
Formula Weight 42.39
Concentration or Composition (by Analyte or Components) 1 M in ethanol
Packaging Glass Bottle
Appearance Clear Liquid
Decomposition Information Carbon Oxides; Hydrogen Chloride Gas; Lithium Oxides
Density 0.83 g/mL (at 20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.