Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chromate, Dihydrate, Spectrum™ Chemical
SDP

Catalog No. 18611310 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-310 125 g Amber Glass Bottle
18-611-311 500 g Amber Glass Bottle
18-611-312 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-310 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1111125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7789-01-7

Specifications

Color Yellow
CAS Max % 1
Physical Form Crystalline Powder
Quantity 125 g
Assay Percent Range 96%
Molecular Formula Li2CrO4·2H2O
Linear Formula Li2CrO4⋅2H2O
UN Number UN1479
Solubility Information Very soluble in water
IUPAC Name dilithium chromate dihydrate
Molecular Weight (g/mol) 165.92
Formula Weight 165.92
Percent Purity ≥96%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Amber Glass Bottle
Sensitivity Deliquescent, Moisture sensitive
Appearance Yellow Crystalline Powder
Density 2.15 g/cm³
Chemical Name or Material Lithium Chromate
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.