Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chromate, Dihydrate, Spectrum™ Chemical
SDP

Catalog No. 18611312 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-312 2.5 kg Amber Glass Bottle
18-611-310 125 g Amber Glass Bottle
18-611-311 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-312 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L11112.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent grade lithium chromate dihydrate, a crystalline powder for laboratory use.

  • Molecular Formula: Li2CrO4·2H2O
  • Molecular Weight: 165.92 g/mol
  • Appearance: Crystalline powder
  • Color: Yellow
  • Odor: Odorless
  • Assay: 96% (MIN)
  • Solubility: Very soluble in water
  • Density: 2.15 g/cm3
  • Decomposition Temperature: 75°C (167°F)
  • Storage: Deliquescent; keep tightly closed

Chemical Identifiers

CAS 7789-01-7

Specifications

Color Yellow
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Molecular Formula Li2CrO4·2H2O
Linear Formula Li2CrO4·2H2O
UN Number UN1479
Solubility Information Very soluble in water
IUPAC Name lithium chromate dihydrate
Molecular Weight (g/mol) 165.92
Formula Weight 165.92
Percent Purity 96%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Amber Glass Bottle
Sensitivity Moisture sensitive, Hygroscopic
Appearance Yellow Crystalline Powder
Density 2.15 g/cm³
Chemical Name or Material Lithium Chromate, Dihydrate
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.