Learn More
Apexbio Technology LLC Lithium Citrate 6080-58-6 500g
SDP
Catalog No. 5000061657
Encompass_Preferred
Lithium Citrate is a lithium-based compound widely utilized in biomedical research due to its ability to modulate mood stability and neuronal signaling Studies indicate that Lithium Citrate influences numerous biochemical pathways including enzyme inhibition and neurotransmitter regulation contributing to mood stabilization effects seen in bipolar disorder (BD) models Experimentally Lithium Citrate has shown capability in reducing self-injurious behaviors and addressing acute manic episodes in BD research models Additionally it facilitates the dissolution of uric acid crystals proposing potential relevance in gout-associated investigations Lithium Citrate also displays inhibitory properties towards prostatic acid phosphatase (PAP) a mechanism behind some observed side effects Currently detailed IC50 data for Lithium Citrate s enzyme inhibition effects remain limited in literature
Catalog No. 50-000-61657 Supplier Apexbio Technology LLC Supplier No. A8462500000
Lithium Citrate is a lithium-based compound widely utilized in biomedical research due to its ability to modulate mood stability and neuronal signaling Studies indicate that Lithium Citrate influences numerous biochemical pathways including enzyme inhibition and neurotransmitter regulation contributing to mood stabilization effects seen in bipolar disorder (BD) models Experimentally Lithium Citrate has shown capability in reducing self-injurious behaviors and addressing acute manic episodes in BD research models Additionally it facilitates the dissolution of uric acid crystals proposing potential relevance in gout-associated investigations Lithium Citrate also displays inhibitory properties towards prostatic acid phosphatase (PAP) a mechanism behind some observed side effects Currently detailed IC50 data for Lithium Citrate s enzyme inhibition effects remain limited in literature