Learn More
Description
- Molecular Formula: C6H5Li3O7·4H2O
- Appearance: White powder or fine crystals
- Melting Point: 333°C
- Water Solubility: Soluble
- Relative Density: 1.64
- Grade: Reagent
Specifications
Specifications
| Melting Point | 333°C |
| Color | White |
| pH | 8.0 to 9.0 |
| CAS Max % | 1 |
| Physical Form | Powder, Needles or Crystals |
| Quantity | 500 g |
| Assay Percent Range | 90% |
| Molecular Formula | C6H5Li3O7·4H2O |
| UN Number | NONH for all modes of transport |
| Synonym | trilithium citrate |
| Show More |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
