Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium cobalt(III) oxide

Catalog No. 442704100GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-270-4100GA 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-270-4100GA Supplier Sigma Aldrich Supplier No. 442704100GA
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Lithium cobalt(III) oxide (LiCoO2) can be used as a cathode material with a specific capacity of ∼274 mAhg1 for the fabrication of lithium-ion batteries. Commercially, these LiCoO2 fabricated Li-ion batteries can be used in a majority of smartphones. LiCoO2 can also be used in the formation of fuel cells.

Specifications

CAS 12190-79-3
Molecular Formula CoLiO2
Linear Formula LiCoO2
MDL Number MFCD00049786
Quantity 100 g
Synonym Lithium cobaltite
Molecular Weight (g/mol) 97.87
Percent Purity 99.8% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.