Learn More
Strem, An Ascensus Company CAS# 12190-79-3. 50g. Lithium cobalt(III) oxide, min. 98%. MFCD00049786
SDP
Catalog No. 5090113688
Encompass
CAS# 12190-79-3. 50g. Lithium cobalt(III) oxide, min. 98%. MFCD00049786. Molecular Weight: 97.87. Molecular Formula: LiCoO2. Color/form: dark gray pwdr. Strem# 93-0386. http://www.strem.com/catalog/v/93-0386/
Catalog No. 50-901-13688 Supplier Strem, An Ascensus Company Supplier No. 93038650G
CAS# 12190-79-3. 50g. Lithium cobalt(III) oxide, min. 98%. MFCD00049786. Molecular Weight: 97.87. Molecular Formula: LiCoO2. Color/form: dark gray pwdr. Strem# 93-0386. http://www.strem.com/catalog/v/93-0386/