Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium cobalt(III) oxide, 99.5% (metals basis)

Catalog No. aa4209014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4209014 25 g
AA4209022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4209014 Supplier Thermo Scientific Chemicals Supplier No. 04209014
Only null left
Add to Cart

CAS: 12190-79-3 | CoLiO2 | 97.871 g/mol

Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12190-79-3
Molecular Formula CoLiO2
Molecular Weight (g/mol) 97.871
MDL Number MFCD00049786
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
PubChem CID 23670860
IUPAC Name lithium;oxido(oxo)cobalt
SMILES [Li+].[O-][Co]=O

Specifications

Color Blue
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 97.88
Odor Odorless
Sensitivity Hygroscopic
Appearance Dark blue powder
Chemical Name or Material Lithium cobalt(III) oxide
Hazard Category H350-H361f
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 235-362-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.