Learn More
Lithium cobalt(III) oxide, 99.5% (metals basis)
Description
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Color | Blue |
| Physical Form | Powder |
| Quantity | 100 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Dark blue powder |
| Assay Percent Range | 99.5% (Metal basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 97.88 |
| Chemical Name or Material | Lithium cobalt(III) oxide |
Safety and Handling
| Hazard Category | H350-H361f |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 235-362-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.