Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium cobalt phosphate

Catalog No. 72514525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-514-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-514-525G Supplier Sigma Aldrich Supplier No. 72514525G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.LCP belongs to the class of orthorhombic antiferromagnetic materials that is majorly used as a high voltage cathode. It can be synthesized by hydrothermal methods and utilized in the fabrication of Li-Ion batteries.Product of Engi-Mat Co.

Specifications

CAS 13824-63-0
Linear Formula LiCoPO4
MDL Number MFCD16878981
Quantity 25 g
Synonym Cobalt lithium phosphate; LCP
Percent Purity ≥99% (trace metals analysis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.