Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Cobaltite, LiCoO2, MP Biomedicals

Catalog No. ICN221085 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN221085 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN221085 Supplier MP Biomedicals, Inc Supplier No. 0522108580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Molecular formula: CoLiO2
  • Molecular weight: 97.871 g/mol
  • CAS number: 12190-79-3
  • Synonym: Lithium cobaltite
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12190-79-3
Molecular Formula CoLiO2
Molecular Weight (g/mol) 97.871
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
PubChem CID 23670860
IUPAC Name lithium;oxido(oxo)cobalt
SMILES [Li+].[O-][Co]=O

Specifications

Quantity 100 g
Linear Formula LiCoO2
Solubility Information Insoluble in water
InChI Formula InChI=1S/Co.Li.2O/q;+1;;-1
Formula Weight 97.871
Appearance Gray Powder
Chemical Name or Material Lithium Cobaltite
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.