Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Diisopropylamide, 2 M Solution in Tetrahydrofuran / Heptane / Ethylbenzene, Spectrum™ Chemical
SDP

Catalog No. 18603934 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-934 500 mL Amber Glass Bottle
18-603-933 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-603-934 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L2005500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L2005, 4111-54-0, C6H14LiN

Spectrum™ Chemical Lithium Diisopropylamide, 2 M Solution in Tetrahydrofuran / Heptane / Ethylbenzene , also known as LDA, is used as a strong base. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Specifications

Chemical Name or Material Lithium Diisopropylamide, 2 M Solution in Tetrahydrofuran / Heptane / Ethylbenzene
Color Reddish-orange, Yellow
CAS 142-82-5, 109-99-9
CAS Max % 29%, 26%
Physical Form Liquid
Quantity 500 mL
Molecular Formula C6H14LiN
Linear Formula [(CH3)2CH]2NLi
UN Number UN3399
Flash Point 2°C (35.6°F)
SMILES CC(C)[N-][Li+]
IUPAC Name lithium N-propan-2-ylpropan-2-amine
Formula Weight 107.08
Grade Ungraded
Concentration or Composition (by Analyte or Components) Approx. 2 M
Manufacturing Origin USA
Packaging Amber Glass Bottle
Sensitivity Moisture sensitive
Appearance Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.