Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Dimethylamide

Catalog No. 29606650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-606-650G 50 g
29-606-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-606-650G Supplier Sigma Aldrich Supplier No. 29606650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium dimethylamide is used as a reagent in the synthesis of boron-nitrogen heterocycles and 2,2'sa?diborabiphenyl. It is a deprotecting agent which is employed in demethylation of bis(imino)pyridine ferrous dichloride for synthesizing ferrous amide-ate complexes.

Specifications

CAS 3585-33-9
Molecular Formula C2H6LiN
Linear Formula (CH3)2NLi
MDL Number MFCD00008295
Quantity 50 g
Molecular Weight (g/mol) 51.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.