Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Dimethylaminoborohydride Solution

Catalog No. 658235100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-823-5100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 65-823-5100ML Supplier Sigma Aldrich Supplier No. 658235100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium dimethylaminoborohydride is a reagent exhibiting dual properties like a metal hydride and nitrogen nucleophile. It is generally considered as an alternative to lithium aluminum hydride (LAH) for the reduction of carbonyl compounds, amides, and lactams.Lithium Aminoborohydride (LAB) ReagentsReactant for:B-H oxidative addition reactionsReduction and amination reactionsReduction of N-alkyl lactamsSynthesis of tertiary amine-boranes

Specifications

CAS 18419-84-6
Density 0.836 g/mL at 25°C
Boiling Point 84°C (50 mmHg)
Molecular Formula C8H22Si2O2
Refractive Index n20/D 1.423
Linear Formula ((CH3CH2O)(CH3)2Si)2
MDL Number MFCD08276886
Quantity 1 g
Synonym N-Methylmethanamine boron complex; Lithium (dimethylamino)trihydroborate; Lithium trihydro(N-methylmethanaminato)borate
Molecular Weight (g/mol) 206.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.