Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Diphenylphosphide Solution

Catalog No. 464872100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-487-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 46-487-2100ML Supplier Sigma Aldrich Supplier No. 464872100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium Diphenylphosphide Solution

Specifications

CAS 2899-29-8
Melting Point 73°C to 77°C
Molecular Formula C11H14N2O
Linear Formula C11H14N2O
MDL Number MFCD00037970
Quantity 100 mL
Synonym Diphenyl lithium phosphide; Diphenylphosphine lithium salt; Diphenylphosphino)lithium; Lithiodiphenyl phosphide; Lithiodiphenylphosphine
Molecular Weight (g/mol) 190.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.