Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium DL-2-Hydroxybutyrate 98.0+%, TCI America™
SDP

Catalog No. H05805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05805G 5 g
H058025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05805G Supplier TCI America Supplier No. H05805G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 381716-41-2
Molecular Formula C4H7LiO3
Molecular Weight (g/mol) 110.037
MDL Number MFCD00040991
InChI Key CILZBARDXHWUSX-UHFFFAOYSA-M
Synonym DL-2-Hydroxybutyric Acid Lithium Salt
PubChem CID 23707041
IUPAC Name lithium;2-hydroxybutanoate
SMILES [Li+].CCC(C(=O)[O-])O

Specifications

Color White
Quantity 5 g
Formula Weight 110.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium DL-2-Hydroxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.