Learn More
Medchemexpress LLC Lithium dodecyl sulf 5g
SDP
Catalog No. 5000206856
Encompass_Preferred
Lithium dodecyl sulfate is an anionic detergent and surfactant that can be used in place of SDS for electrophoresis under low temperatures Lithium dodecyl sulfate can be used for synthesis of nanomaterials and chromatography[1]
Catalog No. 50-002-06856 Supplier Medchemexpress LLC Supplier No. HYW0995355G
Lithium dodecyl sulfate is an anionic detergent and surfactant that can be used in place of SDS for electrophoresis under low temperatures Lithium dodecyl sulfate can be used for synthesis of nanomaterials and chromatography[1]