Learn More
Abcam Lithium dodecyl sulfate, Anionic detergent, 25G
Catalog No. 502704664
Encompass_Preferred
MW 272.3 g/mol. Anionic detergent that may be used for denaturing native proteins in place of SDS (sodium dodecyl sulfate) for gel electrophoresis in cold conditions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4664 Supplier Abcam Supplier No. AB14639025G
MW 272.3 g/mol. Anionic detergent that may be used for denaturing native proteins in place of SDS (sodium dodecyl sulfate) for gel electrophoresis in cold conditions.
The product is subject to the following: Abcam Restricted Use Statement