Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Dodecyl Sulfate, Ultra Pure, MP Biomedicals

Catalog No. ICN800753 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN800753 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN800753 Supplier MP Biomedicals, Inc Supplier No. 04800753
Add to Cart
Add to Cart

2044-56-6 (LDS Lauryl sulfate lithium salt) Purity: >>>99% Exhibits greater solubility than SDS at lower temperatures, while maintaining similar detergency and wetting ability

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
PubChem CID 2735071
IUPAC Name lithium(1+) dodecyl sulfate
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 25 g
Grade Ultra Pure
Chemical Name or Material Lithium Dodecyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.