Learn More
Pickering Laboratories Inc LITHIUM ELUANT 2.75 4X950ML
SDP
Catalog No. NC9745951
Encompass_Preferred
Lithium Eluant, pH 2.75, case of 4 (950 mL/bottle) Amino Acid Analysis.
Catalog No. NC9745951 Supplier Pickering Laboratories Inc Supplier No. LI275
Lithium Eluant, pH 2.75, case of 4 (950 mL/bottle) Amino Acid Analysis.