Learn More
Grainger Sta-Lube Super White Multi-Purpose Lithium Grease, 1.5 NLGI, 14 oz Cartridge
Catalog No. 502768944
Encompass_Preferred
This Super White multi-purpose lithium grease is a semi-solid, white lubricant designed for applications where appearance and cleanliness are essential. Formulated with a lithium-12 base, it provides superior lubrication and durability across a range of environmental conditions. This product contains advanced anti-wear and anti-friction additives to enhance performance and inhibit oxidation.
  • NLGI grade 1.5 consistency
  • Operating temperature range of 0°F to 300°F (−17.8°C to 148.9°C)
  • NSF H2 registered for use in meat and poultry plants where there is no possibility of food contact
  • Excellent water resistance with 10% water washout
  • Neutralizing action on acids provides added protection
  • Convenient 14 oz cartridge container
Catalog No. 50-276-8944 Supplier Grainger Supplier No. 13P453
This Super White multi-purpose lithium grease is a semi-solid, white lubricant designed for applications where appearance and cleanliness are essential. Formulated with a lithium-12 base, it provides superior lubrication and durability across a range of environmental conditions. This product contains advanced anti-wear and anti-friction additives to enhance performance and inhibit oxidation.
  • NLGI grade 1.5 consistency
  • Operating temperature range of 0°F to 300°F (−17.8°C to 148.9°C)
  • NSF H2 registered for use in meat and poultry plants where there is no possibility of food contact
  • Excellent water resistance with 10% water washout
  • Neutralizing action on acids provides added protection
  • Convenient 14 oz cartridge container