Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium hexafluorophosphate solution

Catalog No. 901685500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-168-5500ML 500 mL
90-168-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-168-5500ML Supplier Sigma Aldrich Supplier No. 901685500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium hexafluorophosphate solution in ethylene carbonate, dimethyl carbonate, and diethyl carbonate is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.The solutions of LiPF6 in alkyl carbonates find applications as electrolytes in lithium ion batteries and the use of high quality battery grade electrolytes having extremely low water (<15 PPM) and hydrogen fluoride (<50 PPM) contents are critical for achieving high electrochemical performance.Product of Ube Industries Ltd.

Specifications

Linear Formula LiPF6
Quantity 500 mL
MDL Number MFCD00011096
Synonym 1.0 M LiPF6-»EC/DMC/DEC; 1 M LiPF6 [EC/DMC/DEC=1:1:1 (v/v/v)]; POWERLYTE#EC/DMC/DEC
Biological Activity Kidney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.